Learn More
Medchemexpress LLC 5-(2-(4-fluorophenyl)acetyl)-2,2-dimethyl-1,3-dioxane-4,6-dione | 1174047-11-0 | 98.0% | 280.25 g/mol | C14H13FO5 | 5 G
SDP
Catalog No. 5000285176
Encompass_Preferred
5-(2-(4-Fluorophenyl)acetyl)-2,2-dimethyl-1,3-dioxane-4,6-dione is a Meldrum's acid derivative used as a synthetic intermediate in medicinal chemistry and drug discovery. Supplied as a solid for research use, it supports a range of acylation and derivatization reactions and is offered in multiple package sizes to accommodate small- and large-scale synthesis needs.
  • Used as a drug synthesis intermediate for preparation of active compounds.
  • Meldrum's acid derivative reactivity enables acylation and ring-opening transformations.
  • Supplied as a solid for convenient handling and storage.
  • Available in multiple package sizes to support scale-up.
  • Characterized by analytical data (LC/MS, NMR, HPLC) to support product quality.
Catalog No. 50-002-85176 Supplier Medchemexpress LLC Supplier No. HYW3823605G
5-(2-(4-Fluorophenyl)acetyl)-2,2-dimethyl-1,3-dioxane-4,6-dione is a Meldrum's acid derivative used as a synthetic intermediate in medicinal chemistry and drug discovery. Supplied as a solid for research use, it supports a range of acylation and derivatization reactions and is offered in multiple package sizes to accommodate small- and large-scale synthesis needs.
  • Used as a drug synthesis intermediate for preparation of active compounds.
  • Meldrum's acid derivative reactivity enables acylation and ring-opening transformations.
  • Supplied as a solid for convenient handling and storage.
  • Available in multiple package sizes to support scale-up.
  • Characterized by analytical data (LC/MS, NMR, HPLC) to support product quality.