Learn More
Medchemexpress LLC 5-(2-(4-fluorophenyl)acetyl)-2,2-dimethyl-1,3-dioxane-4,6-dione | 1174047-11-0 | MFCD16876922 | 98.0% | 280.25 g·mol-1 | C14H13FO5 | 1 G
SDP
Catalog No. 5000285156
Encompass_Preferred
5-(2-(4-Fluorophenyl)acetyl)-2,2-dimethyl-1,3-dioxane-4,6-dione is a Meldrum's-acid-derived drug intermediate used for the synthesis of various active pharmaceutical compounds. It is supplied in laboratory-scale quantities and is characterized by its molecular formula C14H13FO5, molecular weight 280.25 g·mol-1, and CAS number 1174047-11-0. Typical reported purity for packaged quantities is 98%.
  • used as a versatile building block in organic synthesis.
  • available in multiple laboratory-scale package sizes, including 1 g.
  • high reported purity supports reliable downstream reactions.
  • well defined molecular formula and molecular weight for formulation and calculation.
  • suitable for research and development applications in medicinal chemistry.
Catalog No. 50-002-85156 Supplier Medchemexpress LLC Supplier No. HYW3823601G
5-(2-(4-Fluorophenyl)acetyl)-2,2-dimethyl-1,3-dioxane-4,6-dione is a Meldrum's-acid-derived drug intermediate used for the synthesis of various active pharmaceutical compounds. It is supplied in laboratory-scale quantities and is characterized by its molecular formula C14H13FO5, molecular weight 280.25 g·mol-1, and CAS number 1174047-11-0. Typical reported purity for packaged quantities is 98%.
  • used as a versatile building block in organic synthesis.
  • available in multiple laboratory-scale package sizes, including 1 g.
  • high reported purity supports reliable downstream reactions.
  • well defined molecular formula and molecular weight for formulation and calculation.
  • suitable for research and development applications in medicinal chemistry.