Learn More
Medchemexpress LLC 5-(2-(4-fluorophenyl)acetyl)-2,2-dimethyl-1,3-dioxane-4,6-dione | 1174047-11-0 | MFCD16876922 | 98.0% | 280.25 g/mol | C14H13FO5 | 10 G
SDP
Catalog No. 5000285189
Encompass_Preferred
5-(2-(4-fluorophenyl)acetyl)-2,2-dimethyl-1,3-dioxane-4,6-dione is a chemical intermediate used in medicinal chemistry for the synthesis of active compounds. It is provided in research-grade quantities for laboratory synthesis and development work.
  • High purity 98.0% suitable for synthetic applications.
  • Available in multiple pack sizes, including 10 G for scale-up.
  • Molecular weight 280.25 g/mol and formula C14H13FO5 for formulation calculations.
  • Stable at room temperature under inert atmosphere; in solvent, store at -80°C (2 years) or -20°C (1 year).
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-85189 Supplier Medchemexpress LLC Supplier No. HYW38236010G
5-(2-(4-fluorophenyl)acetyl)-2,2-dimethyl-1,3-dioxane-4,6-dione is a chemical intermediate used in medicinal chemistry for the synthesis of active compounds. It is provided in research-grade quantities for laboratory synthesis and development work.
  • High purity 98.0% suitable for synthetic applications.
  • Available in multiple pack sizes, including 10 G for scale-up.
  • Molecular weight 280.25 g/mol and formula C14H13FO5 for formulation calculations.
  • Stable at room temperature under inert atmosphere; in solvent, store at -80°C (2 years) or -20°C (1 year).
  • Intended for research use only; not for human or clinical use.