Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-(2-(4-fluorophenyl)acetyl)-2,2-dimethyl-1,3-dioxane-4,6-dione | 1174047-11-0 | MFCD16876922 | 98.0% | 280.25 g/mol | C14H13FO5 | 100 MG
SDP

Catalog No. 5000285241
Encompass_Preferred

5-(2-(4-fluorophenyl)acetyl)-2,2-dimethyl-1,3-dioxane-4,6-dione is a Meldrum's-acid derivative used as a drug intermediate for synthesis of various active compounds. It is supplied for research use in laboratory-scale quantities; molecular formula C14H13FO5, molecular weight 280.25 g/mol, CAS 1174047-11-0.

  • Used as a drug intermediate in organic synthesis.
  • Molecular formula C14H13FO5 and molecular weight 280.25 g/mol.
  • CAS number 1174047-11-0 for precise identification.
  • Available in laboratory-scale package sizes, including 100 mg.
  • Storage recommendations: store under nitrogen at room temperature; in solvent store at -80°C up to 2 years or -20°C up to 1 year.

Catalog No. 50-002-85241 Supplier Medchemexpress LLC Supplier No. HYW382360100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-(2-(4-fluorophenyl)acetyl)-2,2-dimethyl-1,3-dioxane-4,6-dione is a Meldrum's-acid derivative used as a drug intermediate for synthesis of various active compounds. It is supplied for research use in laboratory-scale quantities; molecular formula C14H13FO5, molecular weight 280.25 g/mol, CAS 1174047-11-0.

  • Used as a drug intermediate in organic synthesis.
  • Molecular formula C14H13FO5 and molecular weight 280.25 g/mol.
  • CAS number 1174047-11-0 for precise identification.
  • Available in laboratory-scale package sizes, including 100 mg.
  • Storage recommendations: store under nitrogen at room temperature; in solvent store at -80°C up to 2 years or -20°C up to 1 year.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.