Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-(2-(4-fluorophenyl)acetyl)-2,2-dimethyl-1,3-dioxane-4,6-dione | 1174047-11-0 | MFCD16876922 | 98.0% | 280.25 g/mol | C14H13FO5 | 250 MG
SDP

Catalog No. 5000285044
Encompass_Preferred

5-(2-(4-fluorophenyl)acetyl)-2,2-dimethyl-1,3-dioxane-4,6-dione is a Meldrum's acid derivative used as a drug intermediate for the synthesis of various active compounds. The compound has formula C14H13FO5, CAS number 1174047-11-0, and a molecular weight of 280.25 g/mol. It is offered in multiple pack sizes for research and synthetic applications.

  • Meldrum's acid derivative suitable for organic synthesis
  • Used as a drug intermediate in medicinal chemistry
  • Chemical formula C14H13FO5 and molecular weight 280.25 g/mol
  • CAS number 1174047-11-0 for unambiguous identification
  • High purity (98.0%) suitable for synthetic use
  • Available in multiple pack sizes including 250 mg

Catalog No. 50-002-85044 Supplier Medchemexpress LLC Supplier No. HYW382360250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-(2-(4-fluorophenyl)acetyl)-2,2-dimethyl-1,3-dioxane-4,6-dione is a Meldrum's acid derivative used as a drug intermediate for the synthesis of various active compounds. The compound has formula C14H13FO5, CAS number 1174047-11-0, and a molecular weight of 280.25 g/mol. It is offered in multiple pack sizes for research and synthetic applications.

  • Meldrum's acid derivative suitable for organic synthesis
  • Used as a drug intermediate in medicinal chemistry
  • Chemical formula C14H13FO5 and molecular weight 280.25 g/mol
  • CAS number 1174047-11-0 for unambiguous identification
  • High purity (98.0%) suitable for synthetic use
  • Available in multiple pack sizes including 250 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.