Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(2-Amino-4-chloro-5-sulfamoylphenyl)-1H-tetrazole 98.0+%, TCI America™
SDP

Catalog No. A221925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A221925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A221925G Supplier TCI America Supplier No. A221925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 82212-14-4
Molecular Formula C7H7ClN6O2S
Molecular Weight (g/mol) 274.683
InChI Key OKHJHVJXPHYALI-UHFFFAOYSA-N
Synonym 4-Amino-2-chloro-5-(1H-tetrazol-5-yl)benzenesulfonamide
PubChem CID 10084756
IUPAC Name 4-amino-2-chloro-5-(2H-tetrazol-5-yl)benzenesulfonamide
SMILES C1=C(C(=CC(=C1S(=O)(=O)N)Cl)N)C2=NNN=N2

Specifications

Quantity 25 g
Formula Weight 274.68
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-(2-Amino-4-chloro-5-sulfamoylphenyl)-1H-tetrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.