Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(2-Bromophenyl)-1H-tetrazole 98.0+%, TCI America™
SDP

Catalog No. B25775G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B25775G 5 g
B257725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B25775G Supplier TCI America Supplier No. B25775G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 73096-42-1
Molecular Formula C7H5BrN4
Molecular Weight (g/mol) 225.049
MDL Number MFCD00151799
InChI Key YHVBXKTXLJTDRI-UHFFFAOYSA-N
PubChem CID 560689
IUPAC Name 5-(2-bromophenyl)-2H-tetrazole
SMILES C1=CC=C(C(=C1)C2=NNN=N2)Br

Specifications

Melting Point 182°C
Color White-Yellow
Quantity 5 g
Synonym 5-2-bromophenyl-1h-tetrazole, 5-2-bromophenyl tetrazole, 5-2-bromo-phenyl-2h-tetrazole, 5-2-bromophenyl-2h-tetrazole, 5-2-bromophenyl-1h-1,2,3,4-tetrazole, 5-2-bromophenyl-2h-1,2,3,4-tetrazole, 5-2-bromophenyl-1h-tetraazole, 5-2-bromophenyl-1 h-tetrazole, 1h-tetrazole, 5-2-bromophenyl
Formula Weight 225.05
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(2-Bromophenyl)-1H-tetrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.