Learn More
Aobchem 5-(2-Chloro-3-(trifluoromethyl)phenyl)thiazol-2-amine | ≥95% | CAS 2734776-68-0 | C10H6ClF3N2S | 1g
Catalog No. 5000905618
Encompass_Preferred
5-(2-Chloro-3-(trifluoromethyl)phenyl)thiazol-2-amine | ≥95% | CAS 2734776-68-0 | C10H6ClF3N2S | 1g
Catalog No. 50-009-05618 Supplier Aobchem Supplier No. A582841G
5-(2-Chloro-3-(trifluoromethyl)phenyl)thiazol-2-amine | ≥95% | CAS 2734776-68-0 | C10H6ClF3N2S | 1g