Learn More
Medchemexpress LLC 5-(2-chloroacetamide) thalidomide | 1957235-51-6 | 95.0% | 349.73 g·mol⁻¹ | C15H12ClN3O5 | 100 MG
SDP
Catalog No. 5000284497
Encompass_Preferred
5-(2-chloroacetamide) thalidomide is a thalidomide analogue that incorporates a thiol-reactive chloroacetamide moiety and is used as an E3 ligase-binding ligand for biochemical and cellular research. The compound is supplied as a solid with reported purity of ≥95.0% and a molecular weight of 349.73 g·mol⁻¹.
  • Contains a thiol-reactive chloroacetamide group suitable for covalent target engagement.
  • Useful as an E3 ligase ligand for targeted protein degradation studies.
  • Supplied as a solid form appropriate for laboratory handling and formulation.
  • Reported purity ≥95.0% supports reproducible experimental results.
  • Available in small-mass packs suitable for screening and method development.
Catalog No. 50-002-84497 Supplier Medchemexpress LLC Supplier No. HYW674381100MG
5-(2-chloroacetamide) thalidomide is a thalidomide analogue that incorporates a thiol-reactive chloroacetamide moiety and is used as an E3 ligase-binding ligand for biochemical and cellular research. The compound is supplied as a solid with reported purity of ≥95.0% and a molecular weight of 349.73 g·mol⁻¹.
  • Contains a thiol-reactive chloroacetamide group suitable for covalent target engagement.
  • Useful as an E3 ligase ligand for targeted protein degradation studies.
  • Supplied as a solid form appropriate for laboratory handling and formulation.
  • Reported purity ≥95.0% supports reproducible experimental results.
  • Available in small-mass packs suitable for screening and method development.