Learn More
Medchemexpress LLC 5-(2-chloroacetamide) thalidomide | 1957235-51-6 | MFCD32633381 | 95.0% | 349.73 g·mol⁻¹ | C15H12ClN3O5 | 50 MG
SDP
Catalog No. 5000284641
Encompass_Preferred
5-(2-Chloroacetamide) thalidomide is a thalidomide analogue that incorporates a thiol-specific chloroacetamide electrophile. It is provided as a white to off-white solid for use in chemical biology and PROTAC research. The compound is characterized by CAS 1957235-51-6, molecular formula C15H12ClN3O5, and is supplied at high purity for laboratory use.
  • Thalidomide analogue with a thiol-reactive chloroacetamide group.
  • High purity suitable for chemical biology and degrader design.
  • White to off-white solid that is easy to handle and weigh.
  • Stable under recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • Available in small research quantities for rapid screening.
Catalog No. 50-002-84641 Supplier Medchemexpress LLC Supplier No. HYW67438150MG
5-(2-Chloroacetamide) thalidomide is a thalidomide analogue that incorporates a thiol-specific chloroacetamide electrophile. It is provided as a white to off-white solid for use in chemical biology and PROTAC research. The compound is characterized by CAS 1957235-51-6, molecular formula C15H12ClN3O5, and is supplied at high purity for laboratory use.
  • Thalidomide analogue with a thiol-reactive chloroacetamide group.
  • High purity suitable for chemical biology and degrader design.
  • White to off-white solid that is easy to handle and weigh.
  • Stable under recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • Available in small research quantities for rapid screening.