Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-(2-Chlorophenyl)-1,3,4-thiadiazol-2-amine

Catalog No. CDS002509 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS002509 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS002509 Supplier Sigma Aldrich Supplier No. CDS00250950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-bromo-3-(trifluoromethoxy)benzene undergoes diels-alder reaction with lithium diisopropylamide (lda) in thf and furan, to yield the corresponding 1,4-dihydro-1,4-epoxy-5- or 6-(trifluoromethoxy)naphthalenes.1-bromo-3-(trifluoromethoxy)benzene may be used in the preparation of 1,2-dehydro-3-(trifluoromethoxy)benzene
  • It may be used in the synthesis of new electronically deficient atropisomeric diphosphine ligand (s)-cf3o-biphep [2,2α-bis(diphenylphosphino)-6,6α-ditrifluoromethoxy-1,1α-biphenyl]

Specifications

Molecular Formula C8H6ClN3S
Linear Formula C8H6ClN3S
MDL Number MFCD00548482
Quantity 5 g
Molecular Weight (g/mol) 211.67
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.