Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(2-Chlorophenyl)-1H-tetrazole 98.0+%, TCI America™
SDP

Catalog No. C28115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28115G 5 g
C28111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28115G Supplier TCI America Supplier No. C28115G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50907-46-5
Molecular Formula C7H5ClN4
Molecular Weight (g/mol) 180.60
MDL Number MFCD00040950
InChI Key PSUIIKIEUATWCZ-UHFFFAOYSA-N
Synonym 5-2-chlorophenyl-1h-tetrazole, 5-2-chlorophenyl-2h-tetrazole, 5-2-chlorophenyl-2h-1,2,3,4-tetrazole, 5-2-chlorophenyl tetrazole, 5-2-chlorophenyl-2h-tetraazole, 2h-tetrazole,5-2-chlorophenyl, 5-2-chlorophenyl-1h-1,2,3,4-tetrazole, 5-2-chlorophenyl-1h-1,2,3,4-tetraazole, ldha inhibitor, 14, 2-chlorophenyl tetrazole
PubChem CID 560735
IUPAC Name 5-(2-chlorophenyl)-2H-1,2,3,4-tetrazole
SMILES ClC1=CC=CC=C1C1=NNN=N1

Specifications

Melting Point 179°C
Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 180.60
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material 5-(2-Chlorophenyl)-1H-tetrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.