Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-((2-(dimethylamino)ethyl)amino)-2-methyl-1,3-benzothiazole-4,7-dione | 477603-18-2 | 95.5% and 99.1% | 100 MG
SDP

Catalog No. 5000234070
Encompass_Preferred

BN-82685 is a quinone-based small-molecule inhibitor of CDC25 phosphatases for research use. It shows nanomolar activity against CDC25 family members and is supplied as a solid for biochemical and cellular studies.

  • Quinone-based inhibitor with nanomolar potency against CDC25A, CDC25B, and CDC25C.
  • Solid, brown to red appearance suitable for handling in dry form.
  • Molecular weight 265.33 g/mol; formula C12H15N3O2S.
  • Available in small research pack sizes, including 100 mg.
  • Store at 4°C and protect from light; in solution, store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-34070 Supplier Medchemexpress LLC Supplier No. HY121065100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BN-82685 is a quinone-based small-molecule inhibitor of CDC25 phosphatases for research use. It shows nanomolar activity against CDC25 family members and is supplied as a solid for biochemical and cellular studies.

  • Quinone-based inhibitor with nanomolar potency against CDC25A, CDC25B, and CDC25C.
  • Solid, brown to red appearance suitable for handling in dry form.
  • Molecular weight 265.33 g/mol; formula C12H15N3O2S.
  • Available in small research pack sizes, including 100 mg.
  • Store at 4°C and protect from light; in solution, store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.