Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-[2-(Ethylamino)ethylcarbamoyl]-2-fluorobenzeneboronic acid, 97%, Thermo Scientific™

Catalog No. AAH54499MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54499MD 250 mg
AAH5449903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH54499MD Supplier Thermo Scientific Chemicals Supplier No. H54499MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2096332-79-3
Molecular Formula C11H16BFN2O3
Molecular Weight (g/mol) 254.07
MDL Number MFCD20040204
InChI Key UBDRCBYERBTPRG-UHFFFAOYSA-N
Synonym 5-2-ethylamino ethylcarbamoyl-2-fluorobenzeneboronic acid
PubChem CID 73995842

Specifications

Quantity 250 mg
Formula Weight 254.07
Percent Purity 97%
Chemical Name or Material 5-[2-(Ethylamino)ethylcarbamoyl]-2-fluorobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.