Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[5-(2-Furyl)-1H-pyrazol-3-yl]methanol, 97%, Thermo Scientific™

Catalog No. CC35009CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC35009CB 250 mg Amber Glass Bottle
CC35009DA 1 g Amber Glass Bottle
CC35009DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC35009CB Supplier Thermo Scientific Chemicals Supplier No. CC35009CB
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 84978-67-6
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.164
InChI Key NBADTAIDQSLLHA-UHFFFAOYSA-N
Synonym 5-2-furyl-1h-pyrazol-3-yl methanol, 5-furan-2-yl-1h-pyrazol-3-yl methanol, 3-furan-2-yl-1h-pyrazol-5-yl methanol, 5-2-furyl pyrazol-3-yl methan-1-ol, 1h-pyrazole-3-methanol,5-2-furanyl, 5-2-furyl-1h-pyrazol-4-yl methanol, 5-fur-2-yl-1h-pyrazol-3-yl methanol, 5-furan-2-yl-2h-pyrazol-3-yl methanol
PubChem CID 7060534
IUPAC Name [3-(furan-2-yl)-1H-pyrazol-5-yl]methanol
SMILES C1=COC(=C1)C2=NNC(=C2)CO

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material [5-(2-Furyl)-1H-pyrazol-3-yl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.