Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-(2-(m-Tolyloxy)ethyl)-1,3,4-thiadiazol-2-amine

Catalog No. OTV000402 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV000402 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV000402 Supplier Sigma Aldrich Supplier No. OTV000402250MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Trans,trans-farnesyl acetate is one of the most odor-active or key compounds of hyuganatsu aroma in hyuganatsu (citrus tamurana hort
  • Ex tanaka) peel oil
  • Asymmetric dihydroxylation of trans,trans-farnesyl acetate using the new sharplessα ligand has been studied

Specifications

Molecular Formula C11H13N3OS
Linear Formula C11H13N3OS
MDL Number MFCD08691879
Quantity 10 g
Molecular Weight (g/mol) 235.31
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.