Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[5-(2-Methyl-1,3-thiazol-4-yl)-3-isoxazolyl]methanol, 97%, Thermo Scientific™

Catalog No. CC14009DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC14009DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC14009DA Supplier Thermo Scientific Chemicals Supplier No. CC14009DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 388088-79-7
Molecular Formula C8H8N2O2S
Molecular Weight (g/mol) 196.224
MDL Number MFCD03086111
InChI Key ANQHCDZMYKOBKX-UHFFFAOYSA-N
Synonym 5-2-methyl-1,3-thiazol-4-yl-3-isoxazolyl methanol, 5-2-methyl-1,3-thiazol-4-yl-1,2-oxazol-3-yl methanol, 5-2-methylthiazol-4-yl isoxazol-3-yl methanol, 5-2-methyl-1,3-thiazol-4-yl isoxazol-3-yl methanol, 5-2-methyl-1,3-thiazol-4-yl isoxazol-3-yl methan-1-ol
PubChem CID 2776410
IUPAC Name [5-(2-methyl-1,3-thiazol-4-yl)-1,2-oxazol-3-yl]methanol
SMILES CC1=NC(=CS1)C2=CC(=NO2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Yes
Quantity 1 g
Formula Weight 196.23
Percent Purity 97%
Chemical Name or Material [5-(2-Methyl-1, 3-thiazol-4-yl)-3-isoxazolyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.