Learn More
Medchemexpress LLC 5-(2-methylpiperazin-1-yl)sulfonylisoquinoline | 84477-87-2 | MFCD00063403 | 100.0% | 291.37 g·mol⁻¹ | C14H17N3O2S | 5 MG
SDP
Catalog No. 5000263188
Encompass_Preferred
Protein kinase inhibitor H-7 is a small-molecule inhibitor of protein kinase C (PKC) and cyclic nucleotide-dependent kinases used in biochemical and cell biology research to probe PKC-dependent signaling. Reported properties include a Ki of 6 μM for PKC, molecular formula C14H17N3O2S, and molecular weight 291.37 g·mol⁻¹. It is supplied in small research pack sizes for laboratory use.
  • Inhibits protein kinase C and cyclic nucleotide-dependent kinases.
  • Reported Ki of 6 μM for PKC.
  • Suitable for biochemical and cell signaling studies.
  • High reported purity for research applications.
  • Available in multiple small package sizes for lab use.
Catalog No. 50-002-63188 Supplier Medchemexpress LLC Supplier No. HY1319005MG
Protein kinase inhibitor H-7 is a small-molecule inhibitor of protein kinase C (PKC) and cyclic nucleotide-dependent kinases used in biochemical and cell biology research to probe PKC-dependent signaling. Reported properties include a Ki of 6 μM for PKC, molecular formula C14H17N3O2S, and molecular weight 291.37 g·mol⁻¹. It is supplied in small research pack sizes for laboratory use.
  • Inhibits protein kinase C and cyclic nucleotide-dependent kinases.
  • Reported Ki of 6 μM for PKC.
  • Suitable for biochemical and cell signaling studies.
  • High reported purity for research applications.
  • Available in multiple small package sizes for lab use.