Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(2-Phenyleth-1-ynyl)nicotinic acid, 97%, Thermo Scientific™

Catalog No. KM08328CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
KM08328CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KM08328CB Supplier Thermo Scientific Chemicals Supplier No. KM08328CB
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 175203-69-7
Molecular Formula C14H9NO2
Molecular Weight (g/mol) 223.231
MDL Number MFCD00174055
InChI Key DXJZBANECHYHRT-UHFFFAOYSA-N
Synonym 5-2-phenyleth-1-ynyl nicotinic acid, 5-2-phenylethynyl pyridine-3-carboxylic acid, 5-phenylethynyl pyridine-3-carboxylic acid, 3-pyridinecarboxylicacid, 5-2-phenylethynyl, maybridge1_004925, 5-phenylethynylnicotinic acid, 5-phenylethynyl-nicotinic acid, 5-phenylethynyl nicotinic acid, 5-phenylethynyl-pyridine-3-carboxylic acid
PubChem CID 2779236
IUPAC Name 5-(2-phenylethynyl)pyridine-3-carboxylic acid
SMILES C1=CC=C(C=C1)C#CC2=CC(=CN=C2)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 223.23
Percent Purity 97%
Chemical Name or Material 5-(2-Phenyleth-1-ynyl)nicotinic acid
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.