Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(2-Phenyleth-1-ynyl)thiophene-2-carbaldehyde, 90%, Thermo Scientific™

Catalog No. KM08144EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
KM08144EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KM08144EA Supplier Thermo Scientific Chemicals Supplier No. KM08144EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17257-10-2
Molecular Formula C13H8OS
Molecular Weight (g/mol) 212.266
MDL Number MFCD00126374
InChI Key YFMUACLZRVJOBK-UHFFFAOYSA-N
Synonym 5-2-phenyleth-1-ynyl thiophene-2-carbaldehyde, 5-phenylethynyl thiophene-2-carbaldehyde, 5-2-phenylethynyl thiophene-2-carbaldehyde, 5-phenylethynyl thiophene-2-carboxaldehyde, 2-thiophenecarboxaldehyde,5-2-phenylethynyl, 5-phenylethynyl-thiophene-2-carbaldehyde, 5-phenylethynyl thiophene-2-carboxylate
PubChem CID 2737140
IUPAC Name 5-(2-phenylethynyl)thiophene-2-carbaldehyde
SMILES C1=CC=C(C=C1)C#CC2=CC=C(S2)C=O

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 212.27
Percent Purity 90%
Chemical Name or Material 5-(2-Phenyleth-1-ynyl)thiophene-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.