Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(2-Pyridinyl)-2-thiophenecarbothioamide, 97%, Thermo Scientific™

Catalog No. CC00077DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC00077DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC00077DA Supplier Thermo Scientific Chemicals Supplier No. CC00077DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 306934-91-8
Molecular Formula C10H8N2S2
Molecular Weight (g/mol) 220.308
MDL Number MFCD02677740
InChI Key VOYHCOULKKDLRP-UHFFFAOYSA-N
Synonym 5-2-pyridinyl-2-thiophenecarbothioamide, 5-pyridin-2-yl thiophene-2-carbothioamide, 2-thiophenecarbothioamide,5-2-pyridinyl, 5-pyridin-2-yl thiophene-2-thiocarboxamide, amino 5-2-pyridyl 2-thienyl methane-1-thione
PubChem CID 2776127
IUPAC Name 5-pyridin-2-ylthiophene-2-carbothioamide
SMILES C1=CC=NC(=C1)C2=CC=C(S2)C(=S)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 220.32
Percent Purity 97%
Chemical Name or Material 5-(2-Pyridinyl)-2-thiophenecarbothioamide
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.