Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC [5-(2-thienyl)-3-isoxazolyl]methanol | 194491-44-6 | MFCD03086162 | 99.9% | 181.21 | C8H7NO2S | 250 MG
SDP

Catalog No. 5000303466
Encompass_Preferred

[5-(2-thienyl)-3-isoxazolyl]methanol is a small-molecule AgrA-DNA binding inhibitor (CAS 194491-44-6) used in research on Staphylococcus aureus virulence and infection mechanisms. It is supplied as a light yellow to brown oil and is intended for biochemical and microbiology research applications.

  • Purity 99.88%.
  • Molecular weight 181.21 g/mol.
  • Chemical formula C8H7NO2S.
  • Appearance oil, light yellow to brown.
  • SMILES OCC1=NOC(C2=CC=CS2)=C1.
  • Acts as an AgrA-DNA binding inhibitor for S. aureus infection research.
  • Available in multiple pack sizes suitable for laboratory studies.

Catalog No. 50-003-03466 Supplier Medchemexpress LLC Supplier No. 250MGHYW029719
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

[5-(2-thienyl)-3-isoxazolyl]methanol is a small-molecule AgrA-DNA binding inhibitor (CAS 194491-44-6) used in research on Staphylococcus aureus virulence and infection mechanisms. It is supplied as a light yellow to brown oil and is intended for biochemical and microbiology research applications.

  • Purity 99.88%.
  • Molecular weight 181.21 g/mol.
  • Chemical formula C8H7NO2S.
  • Appearance oil, light yellow to brown.
  • SMILES OCC1=NOC(C2=CC=CS2)=C1.
  • Acts as an AgrA-DNA binding inhibitor for S. aureus infection research.
  • Available in multiple pack sizes suitable for laboratory studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.