Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC [5-(2-thienyl)-3-isoxazolyl]methanol | 194491-44-6 | MFCD03086162 | 99.9% | 181.21 g/mol | C8H7NO2S | 50 MG
SDP

Catalog No. 5000300239
Encompass_Preferred

[5-(2-thienyl)-3-isoxazolyl]methanol is a small-molecule AgrA-DNA binding inhibitor used in research on Staphylococcus aureus virulence and infection. It is provided as a high-purity solid and as concentrated DMSO solutions for biochemical and cellular assays.

  • High purity (~99.9%).
  • Molecular weight 181.21 g/mol.
  • Chemical formula C8H7NO2S.
  • Available as solid and as 10 mM solution in DMSO.
  • Suitable for biochemical and cellular assays targeting AgrA-DNA interactions.

Catalog No. 50-003-00239 Supplier Medchemexpress LLC Supplier No. HYW02971950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

[5-(2-thienyl)-3-isoxazolyl]methanol is a small-molecule AgrA-DNA binding inhibitor used in research on Staphylococcus aureus virulence and infection. It is provided as a high-purity solid and as concentrated DMSO solutions for biochemical and cellular assays.

  • High purity (~99.9%).
  • Molecular weight 181.21 g/mol.
  • Chemical formula C8H7NO2S.
  • Available as solid and as 10 mM solution in DMSO.
  • Suitable for biochemical and cellular assays targeting AgrA-DNA interactions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.