Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[5-(2-Thienyl)-3-isoxazolyl]methanol, 97%, Thermo Scientific™

Catalog No. CC23109CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC23109CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC23109CB Supplier Thermo Scientific Chemicals Supplier No. CC23109CB
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 194491-44-6
Molecular Formula C8H7NO2S
Molecular Weight (g/mol) 181.209
MDL Number MFCD03086162
InChI Key HUAGDHXVPCSWLD-UHFFFAOYSA-N
Synonym 5-2-thienyl-3-isoxazolyl methanol, 5-thiophen-2-yl isoxazol-3-yl methanol, 5-thiophen-2-yl-1,2-oxazol-3-yl methanol, 3-isoxazolemethanol,5-2-thienyl, 5-thiophen-2-ylisoxazol-3-yl methanol, 3mq, 5-2-thienyl isoxazol-3-yl methanol, 5-2-thienyl isoxazol-3-yl methan-1-ol, 5-thiophen-2-yl isoxazol-3-yl methanol .
PubChem CID 2776547
IUPAC Name (5-thiophen-2-yl-1,2-oxazol-3-yl)methanol
SMILES C1=CSC(=C1)C2=CC(=NO2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 181.21
Percent Purity 97%
Chemical Name or Material [5-(2-Thienyl)-3-isoxazolyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.