Learn More
Medchemexpress LLC 5-(3,3-difluorocyclobutyl)-3-[4-[4-methyl-5-(2-(trifluoromethyl)phenyl)-...]]-1,2,4-oxadiazole | 935273-79-3 | 99.2% | 493.47 g/mol | C24H24F5N5O | 1 ML
SDP
Catalog No. 5000215650
Encompass_Preferred
MK-4101 is a small-molecule Smoothened (SMO) antagonist that inhibits the Hedgehog signaling pathway and is used for in vitro cancer and signaling research. Supplied as a ready-to-use 10 mM solution in DMSO (1 mL), the compound is characterized by a confirmed CAS number and molecular weight and should be stored frozen and aliquoted to preserve activity.
  • Potent SMO antagonist suitable for Hedgehog pathway studies.
  • Ready-to-use 10 mM solution in DMSO for rapid experiment setup.
  • Small 1 mL format ideal for pilot studies and limited-scope assays.
  • Well-characterized identity with CAS 935273-79-3 and confirmed molecular weight.
  • Store at -20 °C and aliquot to avoid multiple freeze-thaw cycles.
Catalog No. 50-002-15650 Supplier Medchemexpress LLC Supplier No. HY10003610MM 1ML
MK-4101 is a small-molecule Smoothened (SMO) antagonist that inhibits the Hedgehog signaling pathway and is used for in vitro cancer and signaling research. Supplied as a ready-to-use 10 mM solution in DMSO (1 mL), the compound is characterized by a confirmed CAS number and molecular weight and should be stored frozen and aliquoted to preserve activity.
  • Potent SMO antagonist suitable for Hedgehog pathway studies.
  • Ready-to-use 10 mM solution in DMSO for rapid experiment setup.
  • Small 1 mL format ideal for pilot studies and limited-scope assays.
  • Well-characterized identity with CAS 935273-79-3 and confirmed molecular weight.
  • Store at -20 °C and aliquot to avoid multiple freeze-thaw cycles.