Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-(3,3-difluorocyclobutyl)-3-[4-[4-methyl-5-[2-(trifluoromethyl)phenyl]-1,2,4-triazol-3-yl] | 935273-79-3 | MFCD30343842 | 99.2% | 493.47 g/mol | C24H24F5N5O | 25 MG
SDP

Catalog No. 5000214935
Encompass_Preferred

MK-4101 is a small-molecule Smoothened (SMO) antagonist that inhibits the hedgehog signaling pathway and demonstrates antiproliferative and pro-apoptotic activity in tumor cell models. It is supplied for research use and is characterized by defined molecular properties and high analytical purity.

  • Demonstrated antiproliferative and pro-apoptotic activity in cancer cell models.
  • High purity (≈99.2%).
  • Molecular weight 493.47 g/mol and formula C24H24F5N5O.
  • Soluble in DMSO (≥50 mg/mL) for preparation of stock solutions.
  • Recommended storage conditions for stability in powder and solution forms.

Catalog No. 50-002-14935 Supplier Medchemexpress LLC Supplier No. HY10003625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MK-4101 is a small-molecule Smoothened (SMO) antagonist that inhibits the hedgehog signaling pathway and demonstrates antiproliferative and pro-apoptotic activity in tumor cell models. It is supplied for research use and is characterized by defined molecular properties and high analytical purity.

  • Demonstrated antiproliferative and pro-apoptotic activity in cancer cell models.
  • High purity (≈99.2%).
  • Molecular weight 493.47 g/mol and formula C24H24F5N5O.
  • Soluble in DMSO (≥50 mg/mL) for preparation of stock solutions.
  • Recommended storage conditions for stability in powder and solution forms.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.