Learn More
Medchemexpress LLC 5-(3,3-difluorocyclobutyl)-3-[4-[4-methyl-5-[2-(trifluoromethyl)phenyl]-4h-1,2,4-triazol-3-yl] | 935273-79-3 | 99.2% | 493.47 g/mol | C24H24F5N5O | 50 MG
SDP
Catalog No. 5000215441
Encompass_Preferred
MK-4101 is a small-molecule Smoothened (SMO) antagonist that inhibits the hedgehog signaling pathway and is provided for research use. It has demonstrated cellular activity in reported assays and is supplied as a solid in multiple pre-weighed quantities.
  • Small-molecule SMO antagonist useful for hedgehog pathway studies.
  • Demonstrated cellular activity (IC50 ≈1.1 μM in 293 cells).
  • Reported cellular activity in mouse cells (IC50 ≈1.5 μM).
  • Supplied with high reported purity (~99.2%) suitable for research.
  • Available in multiple pre-weighed quantities, including 50 mg.
  • Characterized molecular properties (CAS 935273-79-3; formula C24H24F5N5O).
Catalog No. 50-002-15441 Supplier Medchemexpress LLC Supplier No. HY10003650MG
MK-4101 is a small-molecule Smoothened (SMO) antagonist that inhibits the hedgehog signaling pathway and is provided for research use. It has demonstrated cellular activity in reported assays and is supplied as a solid in multiple pre-weighed quantities.
  • Small-molecule SMO antagonist useful for hedgehog pathway studies.
  • Demonstrated cellular activity (IC50 ≈1.1 μM in 293 cells).
  • Reported cellular activity in mouse cells (IC50 ≈1.5 μM).
  • Supplied with high reported purity (~99.2%) suitable for research.
  • Available in multiple pre-weighed quantities, including 50 mg.
  • Characterized molecular properties (CAS 935273-79-3; formula C24H24F5N5O).