Learn More
Medchemexpress LLC 5-(3,3-Difluorocyclobutyl)-3-[4-[4-methyl-5-[2-(trifluoromethyl)phenyl]-4H-1,2,4-triazol-3-yl]bicyc | 935273-79-3 | 99.2% | 493.47 | C24H24F5N5O | 10 MG
SDP
Catalog No. 5000215344
Encompass_Preferred
MK-4101 is a small-molecule Smoothened (SMO) antagonist that inhibits the Hedgehog signaling pathway and is intended for preclinical research applications. It has molecular formula C24H24F5N5O and a molecular weight of 493.47 g/mol, and is used in studies of pathway modulation, apoptosis, and oncology-related biology.
  • Potent Smoothened (SMO) antagonist with cellular activity
  • Inhibits Hedgehog signaling for pathway modulation studies
  • High purity suitable for preclinical assays
  • Available in small-scale weights for laboratory research
  • Soluble in DMSO for preparation of stock solutions
Catalog No. 50-002-15344 Supplier Medchemexpress LLC Supplier No. HY10003610MG
MK-4101 is a small-molecule Smoothened (SMO) antagonist that inhibits the Hedgehog signaling pathway and is intended for preclinical research applications. It has molecular formula C24H24F5N5O and a molecular weight of 493.47 g/mol, and is used in studies of pathway modulation, apoptosis, and oncology-related biology.
  • Potent Smoothened (SMO) antagonist with cellular activity
  • Inhibits Hedgehog signaling for pathway modulation studies
  • High purity suitable for preclinical assays
  • Available in small-scale weights for laboratory research
  • Soluble in DMSO for preparation of stock solutions