Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-[3-[[[(4-carboxyphenyl)methyl]amino]carbonyl]-1-methyl-1H-pyrazol-5-yl]-1H-indole-2-carboxylic acid | 1222173-37-6 | 99.1% | 446.46 g·mol⁻1 | C24H22N4O5 | 1MG
SDP

Catalog No. 5000235072
Encompass_Preferred

BI-4394 is a potent, selective, and orally active small-molecule inhibitor of matrix metalloproteinase-13 (MMP-13) used as a research tool to probe MMP-13 biology in biochemical, cellular, and ex vivo cartilage assays.

  • Potent inhibition of MMP-13 (IC50 = 0.001 μM).
  • High selectivity for MMP-13 over other MMPs (>1,000-fold).
  • Chemical formula C24H22N4O5; molecular weight 446.46 g·mol⁻1.
  • High purity (reported commercial offering around 99.1%).
  • Solid formulation; soluble in DMSO up to 10 mM.
  • Recommended storage at -20 °C; reported stability ≥4 years under recommended conditions.
  • Suitable for in vitro biochemical assays, cell-based studies, and ex vivo cartilage degradation models.

Catalog No. 50-002-35072 Supplier Medchemexpress LLC Supplier No. HY1240291MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI-4394 is a potent, selective, and orally active small-molecule inhibitor of matrix metalloproteinase-13 (MMP-13) used as a research tool to probe MMP-13 biology in biochemical, cellular, and ex vivo cartilage assays.

  • Potent inhibition of MMP-13 (IC50 = 0.001 μM).
  • High selectivity for MMP-13 over other MMPs (>1,000-fold).
  • Chemical formula C24H22N4O5; molecular weight 446.46 g·mol⁻1.
  • High purity (reported commercial offering around 99.1%).
  • Solid formulation; soluble in DMSO up to 10 mM.
  • Recommended storage at -20 °C; reported stability ≥4 years under recommended conditions.
  • Suitable for in vitro biochemical assays, cell-based studies, and ex vivo cartilage degradation models.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.