Learn More
Cayman Chemical 5- 3 4-DIhydroxyphenylV 50mg
SDP
Catalog No. 502913536
Encompass_Preferred
An active metabolite of various polyphenols; an inhibitor of MMP-1 and MMP-2 collagen degradation activity (IC50s = 108 and 106 nM, respectively), MMP-1 and MMP-9 gelatin degradation activity (IC50s = 45.2 and 17.7 nM, respectively), and MMP-2 and MMP-9 elastin degradation activity (IC50s = 61.8 and 89 nM, respectively); scavenges free radicals in a TEAC assay
Catalog No. 50-291-3536 Supplier Cayman Chemical Supplier No. 3669050 MG
An active metabolite of various polyphenols; an inhibitor of MMP-1 and MMP-2 collagen degradation activity (IC50s = 108 and 106 nM, respectively), MMP-1 and MMP-9 gelatin degradation activity (IC50s = 45.2 and 17.7 nM, respectively), and MMP-2 and MMP-9 elastin degradation activity (IC50s = 61.8 and 89 nM, respectively); scavenges free radicals in a TEAC assay