Learn More
Cayman Chemical 5-(3',4'-Dihydroxyphenyl)-γ-Valerolactone, Purity: ≥95%, CAS: 21618-92-8, 5 mg
SDP
Catalog No. 502913251
Encompass_Preferred
An active metabolite of various polyphenols; an inhibitor of MMP-1 and MMP-2 collagen degradation activity (IC50s = 108 and 106 nM, respectively), MMP-1 and MMP-9 gelatin degradation activity (IC50s = 45.2 and 17.7 nM, respectively), and MMP-2 and MMP-9 elastin degradation activity (IC50s = 61.8 and 89 nM, respectively); scavenges free radicals in a TEAC assay
Catalog No. 50-291-3251 Supplier Cayman Chemical Supplier No. 366905 MG
An active metabolite of various polyphenols; an inhibitor of MMP-1 and MMP-2 collagen degradation activity (IC50s = 108 and 106 nM, respectively), MMP-1 and MMP-9 gelatin degradation activity (IC50s = 45.2 and 17.7 nM, respectively), and MMP-2 and MMP-9 elastin degradation activity (IC50s = 61.8 and 89 nM, respectively); scavenges free radicals in a TEAC assay