Learn More
Sklar Instruments Scissors, Joseph Scissors, Sklar 5-3/4 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA.
SDP
Catalog No. 501093761 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Joseph Scissors, Sklar, 5-3/4 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA. Sklar Joseph Scissors are small, delicate scissors used for cutting fine tissues. Available curved or straight, they are commonly used in plastic surgery and ENT procedures. This product is straight with smooth edges, sharp/sharp tips, and a length of 5-3/4 inches.
Catalog No. 50-109-3761 Supplier Sklar Instruments Supplier No. 411050
Scissors, Joseph Scissors, Sklar, 5-3/4 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA. Sklar Joseph Scissors are small, delicate scissors used for cutting fine tissues. Available curved or straight, they are commonly used in plastic surgery and ENT procedures. This product is straight with smooth edges, sharp/sharp tips, and a length of 5-3/4 inches.