Learn More
Medchemexpress LLC 5-[3,5-bis(trifluoromethyl)phenyl]-2H-tetrazole | 175205-09-1 | 99.9% | 10 G

Supplier: Medchemexpress LLC HY12008710G
5-[3,5-bis(trifluoromethyl)phenyl]-2H-tetrazole is a research-grade small molecule inhibitor of HIF signaling that disrupts ARNT/TACC3 interactions. It is supplied as a white to off-white solid with high purity for biochemical and cellular research.
- High purity suitable for research applications.
- Molecular formula C9H4F6N4; molecular weight 282.15 g/mol.
- White to off-white solid appearance.
- Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (6 months), -20°C (1 month).
- Available in multiple pack sizes, including 10 G packaging.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.