Learn More
Ace Glass, Inc. Gasket, PTFE, grooved, 0.8mm thick, 136.7mm OD, for 500mL-1000mLflask
SDP
Catalog No. 509557320
Encompass
0.8mm thick X 136.7mm O.D. grooved PTFE gasket used for sealing two flat ground glass surfaces. For use with 500-1000mL reaction flasks, with either conical or flat-ground flanges. Gasket reduces breakage and makes an effective seal.
Catalog No. 50-955-7320 Supplier Ace Glass, Inc. Supplier No. 649521
0.8mm thick X 136.7mm O.D. grooved PTFE gasket used for sealing two flat ground glass surfaces. For use with 500-1000mL reaction flasks, with either conical or flat-ground flanges. Gasket reduces breakage and makes an effective seal.