Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-(3-bromophenyl)-1,3-dihydro-benzofuro[3,2-e]diazepin-2-one | 768404-03-1 | MFCD18086878 | 99.9% | 355.19 | C17H11BrN2O2 | 10 MG
SDP

Catalog No. 5000248311
Encompass_Preferred

5-BDBD is a potent, selective P2X4 receptor antagonist used for in vitro pharmacology and cellular research. It inhibits rP2X4R-mediated currents (IC50 ~0.75 μM) and is provided as a high-purity solid suitable for preparing DMSO stock solutions and conducting receptor assays.

  • Potent P2X4 receptor antagonist with reported IC50 ~0.75 μM.
  • High purity (≈99.9%).
  • Molecular formula C17H11BrN2O2; molecular weight 355.19 g/mol.
  • Solid, white to light yellow, appropriate for dissolution and storage.
  • Soluble in DMSO at 62.5 mg/mL; may require sonication and heating to 60°C.
  • Recommended storage: powder at -20°C for long term; solutions at -80°C for short term.
  • Supplied in small research-scale quantities for laboratory experiments.

Catalog No. 50-002-48311 Supplier Medchemexpress LLC Supplier No. HY10191110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-BDBD is a potent, selective P2X4 receptor antagonist used for in vitro pharmacology and cellular research. It inhibits rP2X4R-mediated currents (IC50 ~0.75 μM) and is provided as a high-purity solid suitable for preparing DMSO stock solutions and conducting receptor assays.

  • Potent P2X4 receptor antagonist with reported IC50 ~0.75 μM.
  • High purity (≈99.9%).
  • Molecular formula C17H11BrN2O2; molecular weight 355.19 g/mol.
  • Solid, white to light yellow, appropriate for dissolution and storage.
  • Soluble in DMSO at 62.5 mg/mL; may require sonication and heating to 60°C.
  • Recommended storage: powder at -20°C for long term; solutions at -80°C for short term.
  • Supplied in small research-scale quantities for laboratory experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.