Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(3-Bromophenyl)-1H-tetrazole, 97%, Thermo Scientific™

Catalog No. AAL15180MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL15180MD 250 mg
AAL1518003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL15180MD Supplier Thermo Scientific Chemicals Supplier No. L15180MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3440-99-1
Molecular Formula C7H5BrN4
Molecular Weight (g/mol) 225.05
MDL Number MFCD01075675
InChI Key UVKPUDRFBHSFJH-UHFFFAOYSA-N
Synonym 5-3-bromophenyl-1h-tetrazole, 5-3-bromophenyl-2h-tetrazole, 5-3-bromo-phenyl-2h-tetrazole, 2h-tetrazole,5-3-bromophenyl, 2h-tetrazole, 5-3-bromophenyl, 5-3-bromophenyl-1h-1,2,3,4-tetrazole, 5-3-bromophenyl-2h-1,2,3,4-tetrazole, zlchem 110, acmc-209i7d, 5-3-bromophenyl tetrazole
PubChem CID 4073056
IUPAC Name 5-(3-bromophenyl)-2H-tetrazole
SMILES BrC1=CC=CC(=C1)C1=NNN=N1

Specifications

Melting Point 150°C to 152°C
Quantity 250 mg
Beilstein 141258
Formula Weight 225.05
Percent Purity 97%
Chemical Name or Material 5-(3-Bromophenyl)-1H-tetrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.