Learn More
Medchemexpress LLC 5-(3-chloro-4-cyclohexylphenyl)-1-(3-methoxyphenyl)-1H-pyrazole-3-carboxylic acid | 1393814-38-4 | 99.7% | 410.89 | C23H23ClN2O3 | 100 MG
SDP
Catalog No. 5000240092
Encompass_Preferred
TC LPA5 4 is a selective, non-lipid antagonist of the lysophosphatidic acid receptor LPA5 (GPR92). It inhibits LPA-induced platelet aggregation and reduces proliferation and migration in certain cancer cell lines, with reported IC50 ≈ 0.8 μM in LPA5-RH7777 cells. Supplied as a high-purity research compound (C23H23ClN2O3, MW 410.89).
  • Selective LPA5 receptor antagonist suitable for receptor pharmacology studies.
  • Inhibits LPA-induced platelet aggregation with low micromolar potency.
  • High purity for reliable biological assay results.
  • Characterized by molecular formula C23H23ClN2O3 and MW 410.89.
  • Available in milligram-scale packs for preclinical research.
Catalog No. 50-002-40092 Supplier Medchemexpress LLC Supplier No. HY107615100MG
TC LPA5 4 is a selective, non-lipid antagonist of the lysophosphatidic acid receptor LPA5 (GPR92). It inhibits LPA-induced platelet aggregation and reduces proliferation and migration in certain cancer cell lines, with reported IC50 ≈ 0.8 μM in LPA5-RH7777 cells. Supplied as a high-purity research compound (C23H23ClN2O3, MW 410.89).
  • Selective LPA5 receptor antagonist suitable for receptor pharmacology studies.
  • Inhibits LPA-induced platelet aggregation with low micromolar potency.
  • High purity for reliable biological assay results.
  • Characterized by molecular formula C23H23ClN2O3 and MW 410.89.
  • Available in milligram-scale packs for preclinical research.