Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-(3'-Hydroxyphenyl)-γ-valerolactone | 21618-91-7 | 99.9% | 192.21 | 5 MG
SDP

Catalog No. 5000393450
Encompass_Preferred

5-(3'-Hydroxyphenyl)-γ-valerolactone is a flavan-3-ols metabolite of human faecal microbiota.

  • Molecular weight: 192.21
  • Formula: C11H12O3
  • Cas number: 21618-91-7
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 99.9%
  • Structure classification: Phenols, monophenols
  • Initial source: Endogenous metabolite
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder at -20°C for 3 years; in solvent at -80°C for 6 months, and -20°C for 1 month

Catalog No. 50-003-93450 Supplier Medchemexpress LLC Supplier No. 5MGHYW165608
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-(3'-Hydroxyphenyl)-γ-valerolactone is a flavan-3-ols metabolite of human faecal microbiota.

  • Molecular weight: 192.21
  • Formula: C11H12O3
  • Cas number: 21618-91-7
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 99.9%
  • Structure classification: Phenols, monophenols
  • Initial source: Endogenous metabolite
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder at -20°C for 3 years; in solvent at -80°C for 6 months, and -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.