Learn More
JANT PHARMACAL CORPORATION ACCUTEST - 5+3 Drug Test Cup - AMP (1000), COC (300), OPI (2000), MET (1000), THC (50) Plus Adulteration Screen (pH/ S.G./ Oxidant) 25 Tests/Kit
SDP
Catalog No. NC3457077
Encompass_Preferred
The ACCUTEST 5 panel multi drug screen + adulteration screens is an easy, fast, qualitative, visually read competitive binding immunoassay method for screening without the need of instrumentation.  The cup is FDA (510) K Cleared and CLIA Waived.The 5 panel + 3 adulteration drug test cup screens for the presence of Amphetamine, Cocaine, Opiates, Methadone, Marijuana and adulterants (pH, Oxidants, Specific Gravity) in human urine, which is qualitativly analzyed to indicate a positive or negative test result.
Catalog No. NC3457077 Supplier JANT PHARMACAL CORPORATION Supplier No. DS84A
The ACCUTEST 5 panel multi drug screen + adulteration screens is an easy, fast, qualitative, visually read competitive binding immunoassay method for screening without the need of instrumentation.  The cup is FDA (510) K Cleared and CLIA Waived.The 5 panel + 3 adulteration drug test cup screens for the presence of Amphetamine, Cocaine, Opiates, Methadone, Marijuana and adulterants (pH, Oxidants, Specific Gravity) in human urine, which is qualitativly analzyed to indicate a positive or negative test result.