Learn More
AVANTOR MASTERFLEX (VWR LLC) Manual Pinch Valve, 5/32"" to 1/4"" tube OD
Catalog No. NC1352686
Encompass
Fluid contacts only the tubing¿ideal for use in pharmaceutical and food industries where eliminating contamination is critical. Obtain precise and repeatable flow rates using easy-to-read metered scale on valve body. Valves are made of Delrin. Tubing is not included¿order flexible tubing with durometer (Shore A) of 60 or less from our "Tubing" section.
Catalog No. NC1352686 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9800200
Fluid contacts only the tubing¿ideal for use in pharmaceutical and food industries where eliminating contamination is critical. Obtain precise and repeatable flow rates using easy-to-read metered scale on valve body. Valves are made of Delrin. Tubing is not included¿order flexible tubing with durometer (Shore A) of 60 or less from our "Tubing" section.