Learn More
Accuflow Systems Saint-Gobain Silicone Tubing, Platinum-Cured, 0.156 in ID, 0.41 in OD
Catalog No. 502365874
Encompass_Preferred
Sani-Tech STHT-C platinum-cured silicone tubing designed for sanitary biopharmaceutical fluid transfer. The product provides a smooth, ultra-pure bore, good thermal stability, and traceability marking for sizes with ID ≥ 1/16 in.
  • Platinum-cured silicone material, biopharmaceutical grade.
  • Inner diameter 0.156 in (3.97 mm), outer diameter 0.41 in (10.32 mm).
  • Wall thickness 0.13 in (3.18 mm), 25 ft coil standard.
  • Durometer hardness 50 Shore A for the STHT-C formulation.
  • Autoclavable and resistant to adherence of blood and sanitary fluids.
  • Laser marked for part number and lot code traceability on IDs ≥ 1/16 in.
  • Suitable for pharmaceutical and biopharmaceutical fluid transfer and single-use assemblies.
Catalog No. 50-236-5874 Supplier Accuflow Systems Supplier No. STHTC1564
Sani-Tech STHT-C platinum-cured silicone tubing designed for sanitary biopharmaceutical fluid transfer. The product provides a smooth, ultra-pure bore, good thermal stability, and traceability marking for sizes with ID ≥ 1/16 in.
  • Platinum-cured silicone material, biopharmaceutical grade.
  • Inner diameter 0.156 in (3.97 mm), outer diameter 0.41 in (10.32 mm).
  • Wall thickness 0.13 in (3.18 mm), 25 ft coil standard.
  • Durometer hardness 50 Shore A for the STHT-C formulation.
  • Autoclavable and resistant to adherence of blood and sanitary fluids.
  • Laser marked for part number and lot code traceability on IDs ≥ 1/16 in.
  • Suitable for pharmaceutical and biopharmaceutical fluid transfer and single-use assemblies.