Learn More
Cayman Chemical 5-(3-Aminoallyl)uridine-5'-O-triphosphate (sodium salt), Purity: ≥95%, CAS: 85280-66-6, 2.7 mg
SDP
Catalog No. 502918247
Encompass_Preferred
An amine-modified nucleotide; has been used in the synthesis of affinity-tagged UTP; incorporation of aminoallyl-UTP into cRNA and bacterial tmRNA has been used in the generation of fluorescently labeled probes for microarrays
Catalog No. 50-291-8247 Supplier Cayman Chemical Supplier No. 384232.7 MG
An amine-modified nucleotide; has been used in the synthesis of affinity-tagged UTP; incorporation of aminoallyl-UTP into cRNA and bacterial tmRNA has been used in the generation of fluorescently labeled probes for microarrays