Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-[[4-[[(1S)-2-hydroxy-1-phenylethyl]amino]-5-(1,3,4-oxadiazol-2-yl)pyrimidin-2-yl]amino]-3,3-dimeth | 2320462-65-3 | 99.3% | C24H22N6O4 | 10MG
SDP

Catalog No. 5000261841
Encompass_Preferred

HPK1-IN-7 is a potent, orally active inhibitor of hematopoietic progenitor kinase 1 (HPK1, MAP4K1) with a reported IC50 of 2.6 nM. It is provided as a research-grade small-molecule powder of high purity for use in biochemical and cellular studies of HPK1 signaling pathways.

  • Potent HPK1 inhibition with reported IC50 of 2.6 nM.
  • High kinome selectivity for reduced off-target activity.
  • High purity (99.3%) suitable for research use.
  • Molecular formula C24H22N6O4; molecular weight 458.47 g/mol.
  • Supplied as a powder for in vitro and in vivo formulation.
  • Recommended storage at -20°C for long-term stability.

Catalog No. 50-002-61841 Supplier Medchemexpress LLC Supplier No. HY13874210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HPK1-IN-7 is a potent, orally active inhibitor of hematopoietic progenitor kinase 1 (HPK1, MAP4K1) with a reported IC50 of 2.6 nM. It is provided as a research-grade small-molecule powder of high purity for use in biochemical and cellular studies of HPK1 signaling pathways.

  • Potent HPK1 inhibition with reported IC50 of 2.6 nM.
  • High kinome selectivity for reduced off-target activity.
  • High purity (99.3%) suitable for research use.
  • Molecular formula C24H22N6O4; molecular weight 458.47 g/mol.
  • Supplied as a powder for in vitro and in vivo formulation.
  • Recommended storage at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.