Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-[4-(2,6-dihydroxyphenyl)phenyl]-1H-indole-3-carbonitrile | 1179347-65-9 | 98.0% | 326.35 g/mol | C21H14N2O2 | 5 MG
SDP

Catalog No. 5000210907
Encompass_Preferred

MT 63-78 is a small-molecule direct activator of AMP-activated protein kinase (AMPK) with an EC50 of 25 μM. It induces mitotic arrest and apoptosis, and inhibits lipogenesis and the mTORC1 pathway. Supplied as a solid for biochemical and cellular research.

  • Activates AMPK with an EC50 of 25 μM.
  • Induces mitotic arrest and apoptosis in cells.
  • Inhibits lipogenesis and the mTORC1 pathway.
  • Solid, light yellow to brown, purity 98.0%.
  • Soluble in DMSO (125 mg/mL); in vivo formulations available (≥2.08 mg/mL in recommended vehicles).
  • Storage: powder 4°C; in solution -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-10907 Supplier Medchemexpress LLC Supplier No. HYW0588495MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MT 63-78 is a small-molecule direct activator of AMP-activated protein kinase (AMPK) with an EC50 of 25 μM. It induces mitotic arrest and apoptosis, and inhibits lipogenesis and the mTORC1 pathway. Supplied as a solid for biochemical and cellular research.

  • Activates AMPK with an EC50 of 25 μM.
  • Induces mitotic arrest and apoptosis in cells.
  • Inhibits lipogenesis and the mTORC1 pathway.
  • Solid, light yellow to brown, purity 98.0%.
  • Soluble in DMSO (125 mg/mL); in vivo formulations available (≥2.08 mg/mL in recommended vehicles).
  • Storage: powder 4°C; in solution -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.