Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan, 97%, Thermo Scientific™

Catalog No. AC452630010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC452630010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC452630010 Supplier Thermo Scientific Chemicals Supplier No. 452630010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 519054-55-8
Molecular Formula C14H17BO3
Molecular Weight (g/mol) 244.10
MDL Number MFCD04115374
InChI Key ZQCCCOMKYKVFFN-UHFFFAOYSA-N
Synonym 2-benzofuran-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-benzofuran, benzofuran-5-boronic acid pinacol ester, 2-1-benzofuran-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, benzo b furan-5-boronic acid, pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzo b furan, benzofuran, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzofuran, pubchem20151, 5-4,4,5,5-tetramethyl-1,3-dioxaborolan-2-yl-1-benzofuran
PubChem CID 2795190
IUPAC Name 2-(1-benzofuran-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C2OC=CC2=C1

Specifications

Melting Point 72.0°C to 73.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 244.1
Percent Purity 97%
Chemical Name or Material 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation.
Precautionary Statement GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.