Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzofurazan, 97%

Catalog No. AC430470050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430470050 5 g Glass bottle
AC430470010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC430470050 Supplier Thermo Scientific Chemicals Supplier No. 430470050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1073355-14-2 | C12H15BN2O3 | 246.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073355-14-2
Molecular Formula C12H15BN2O3
Molecular Weight (g/mol) 246.07
MDL Number MFCD09842717
InChI Key MQXANAGMQAYTMV-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzo c 1,2,5 oxadiazole, benzofurazan-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,1,3-benzoxadiazole, benzofurazan-5-boronic acid, pinacol ester, benzo c 1,2,5 oxadiazole-5-boronic acid, pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzofurazan, benzo c 1,2,5 oxadiazole-5-boronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,1,3-benzoxadiazole, benzo c 1,2,5 oxadiazol-5-ylboronic acid pinacol ester, amtb001
PubChem CID 46738004
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzoxadiazole
SMILES CC1(C)OB(OC1(C)C)C1=CC2=NON=C2C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 88.0°C to 90.0°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 246.08
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzofurazan
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.