Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. T34855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34855G 5 g
T34851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34855G Supplier TCI America Supplier No. T34855G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1040281-83-1
Molecular Formula C11H15BO3S
Molecular Weight (g/mol) 238.108
MDL Number MFCD09266186
InChI Key OVGYVJMTHQRCDB-UHFFFAOYSA-N
Synonym 2-(5-Formyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-Formyl-2-thiopheneboronic Acid Pinacol Ester
PubChem CID 57415686
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbaldehyde
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(S2)C=O

Specifications

Melting Point 68°C
Color White-Yellow
Quantity 5 g
Formula Weight 238.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.