Learn More
Medchemexpress LLC 5-[(4,5-dimethoxy-2-propan-2-ylphenyl)methyl]pyrimidine-2,4-diamine | 1026582-88-6 | MFCD12828747 | 97.3% | 302.37 g/mol | C16H22N4O2 | 10 MG
SDP
Catalog No. 5000296965
Encompass_Preferred
RO-3 is a small-molecule research compound that selectively antagonizes P2X3 and P2X2/3 purinergic receptors. It is described as CNS-penetrant and orally active, and is supplied as a solid for laboratory research use only. Handle according to safety data and supplier instructions for storage and use.
  • Selective P2X3 and P2X2/3 antagonist.
  • CNS-penetrant and orally active.
  • Reported purity approximately 97.3%.
  • Available in small milligram-scale quantities for research use.
  • Solid form with recommended cold storage for long-term stability.
Catalog No. 50-002-96965 Supplier Medchemexpress LLC Supplier No. HY1997810MG
RO-3 is a small-molecule research compound that selectively antagonizes P2X3 and P2X2/3 purinergic receptors. It is described as CNS-penetrant and orally active, and is supplied as a solid for laboratory research use only. Handle according to safety data and supplier instructions for storage and use.
  • Selective P2X3 and P2X2/3 antagonist.
  • CNS-penetrant and orally active.
  • Reported purity approximately 97.3%.
  • Available in small milligram-scale quantities for research use.
  • Solid form with recommended cold storage for long-term stability.