Learn More
Cayman Chemical 5-(4,6-Dichloro-s-triazin-2-ylamino)fluorescein (hydrochloride), Purity: ≥98%, CAS: 21811-74-5, 50 mg
SDP
Catalog No. 502950760
Encompass_Preferred
An amine-reactive fluorescent protein-labeling agent; has been conjugated to IgG for use as fluorescent antibodies and as a pre-column or on-column reagent to label amino acids and amino acid-containing compounds; has been used as a derivatizing agent in the analysis of ephedrine and pseudoephedrine; ex/em = 492/513 nm, respectively
Catalog No. 50-295-0760 Supplier Cayman Chemical Supplier No. 3408050 MG
An amine-reactive fluorescent protein-labeling agent; has been conjugated to IgG for use as fluorescent antibodies and as a pre-column or on-column reagent to label amino acids and amino acid-containing compounds; has been used as a derivatizing agent in the analysis of ephedrine and pseudoephedrine; ex/em = 492/513 nm, respectively