Learn More
Cayman Chemical 5- 4- AzIdomethyl 1 1-bIph 5mg
SDP
Catalog No. 502915975
Encompass_Preferred
A potential impurity found in commercial preparations of tetrazole-containing sartans
Catalog No. 50-291-5975 Supplier Cayman Chemical Supplier No. 387805 MG
A potential impurity found in commercial preparations of tetrazole-containing sartans